2-Chloro-6-methylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Chloro-2-picoline
B20174-14
B20174-06
CAAAAB20174-14EA
309.68
CAD
CAAAAB20174-14
CAAAB20174-03
CAAAB20174-06
2-Chloro-6-methylpyridine 98%
2-Chloro-6-methylpyridine
Purity: 98%
CAS No.: 18368-63-3
Molecular Formula: C6H6ClN
Formula Weight: 127.57
Flash Point: 73°(163°F)
Refractive Index: 1.5265
Boiling Point: 64-68°/10mm
Beilstein Reference: 107187
UN No.: 2810
MDL No.: MFCD00006245
Formula:
C₆H₆ClN Boiling Pt: 64…68 °C/10 mm Density: 1.167 Flash Pt: 73 °C(163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006245 CAS Number: 18368-63-3 EINECS: 242-241-6 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...