Methyl-2-chloro-4-nitrobenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-4-nitrobenzoic acid methyl ester, Methyl 2-chloro-4-nitrobenzoate

B20169-22 B20169-06 B20169-14
CAAAAB20169-22EA 350.76 CAD
CAAAAB20169-22 CAAAB20169-06 CAAAB20169-14
Methyl-2-chloro-4-nitrobenzoate 98%
Methyl 2-chloro-4-nitrobenzoate

Purity: 98% CAS No.: 13324-11-3 Molecular Formula: C8H6ClNO4 Formula Weight: 215.59 Melting Point: 74-78° Beilstein Reference: 1877723

Formula: C₈H₆ClNO₄
MW: 215.59
Melting Pt: 74…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00017013
CAS Number: 13324-11-3
EINECS: 236-362-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR