1H,1H,2H,2H-Tridecafluoro-1-octanol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctan-1-ol, 1H,1H,2H,2H-Perfluoro-1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanol, 1H,1H,2H,2H-Perfluorooctan-1-ol, 1H,1H,2H,2H-Perfluorooctanol
B20156-30
B20156-09
B20156-18
CAAAAB20156-30EA
2362.1
CAD
CAAAAB20156-30
CAAAB20156-09
CAAAB20156-18
1H,1H,2H,2H-Tridecafluoro-1-octanol 97%
1H,1H,2H,2H-Tridecafluoro-1-octanol
Purity: 97%
CAS No.: 647-42-7
Molecular Formula: CF3(CF2)5(CH2)2OH
Formula Weight: 364.11
Flash Point: 91°(195°F)
Refractive Index: 1.3140
Boiling Point: 90°/20mm
Beilstein Reference: 1992757
MDL No.: MFCD00042143
Formula:
C₈H₅F₁₃O Boiling Pt: 90 °C/20 mm Density: 1.68 Flash Pt: 91 °C(195 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042143 CAS Number: 647-42-7 EINECS: 211-477-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...