(±)-1-Penten-3-ol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(±)-Ethyl vinyl carbinol, Pent-1-en-3-ol
B20155-06
B20155-14
CAAAAB20155-22EA
728.1
CAD
CAAAAB20155-22
CAAAB20155-06
CAAAB20155-14
(±)-1-Penten-3-ol 98+%
(±)-1-Penten-3-ol
Purity: 98+%
CAS No.: 616-25-1
Molecular Formula: C5H10O
Formula Weight: 86.13
Flash Point: 25°(77°F)
Refractive Index: 1.4250
Boiling Point: 114-115°
Beilstein Reference: 1719834
UN No.: 1987
MDL No.: MFCD00004573
Formula:
C₅H₁₀O MW: 86.13 g/mol Boiling Pt: 114…115 °C Density: 0.838 Flash Pt: 25 °C (77 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004573 CAS Number: 616-25-1 EINECS: 210-472-1 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...