1H,1H,7H-Dodecafluoro-1-heptanol 97%

Supplier: Thermo Scientific Chemicals
Warning

B20144-36 B20144-14 B20144-22
CAAAAB20144-36EA 824.76 CAD
CAAAAB20144-36 CAAAB20144-14 CAAAB20144-22
1H,1H,7H-Dodecafluoro-1-heptanol 97%
1H,1H,7H-Dodecafluoro-1-heptanol

Purity: 97% CAS No.: 335-99-9 Molecular Formula: H(CF2)6CH2OH Formula Weight: 332.09 Flash Point: None Refractive Index: 1.3180 Melting Point: -20° Boiling Point: 169-170° Beilstein Reference: 1800110 MDL No.: MFCD00039630

Formula: C₇H₄F₁₂O
Boiling Pt: 169…170 °C
Melting Pt: –20 °C
Density: 1.762
Flash Pt: None
Storage Temperature: Ambient
MDL Number: MFCD00039630
CAS Number: 335-99-9
EINECS: 206-406-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR