[2.2]-Paracyclophane 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Tricyclo[8,2,2,24,7]hexadeca-4,6,10,12,13,15-hexaene, Tricyclo[8.2.2.2(4,7)]hexadeca-1(12),4,6,10,13,15-hexaene

B20136-03 B20136-06
CAAAB20136-03EA 94.48 CAD
CAAAB20136-03 CAAAB20136-06
[2.2]-Paracyclophane 99%
[2.2]-Paracyclophane

Purity: 99% CAS No.: 1633-22-3 Molecular Formula: C16H16 Formula Weight: 208.30 Odor: Odorless Melting Point: 283-288° Beilstein Reference: 1910888 MDL No.: MFCD00003707

Formula: C₁₆H₁₆
MW: 208.3 g/mol
Boiling Pt: 321 °C (1013 hPa)
Melting Pt: 283…288 °C
Density: 1.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003707
CAS Number: 1633-22-3
EINECS: 216-644-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR