Butyl propionate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20125-22
B20125-36
CAAAB20125-22EA
45.98
CAD
CAAAB20125-22
CAAAB20125-36
Butyl propionate 99%
Butyl propionate
Purity: 99%
CAS No.: 590-01-2
Molecular Formula: CH3CH2CO2(CH2)3CH3
Formula Weight: 130.19
Flash Point: 38°(101°F)
Refractive Index: 1.4020
Boiling Point: 144-146°
Merck Reference: 14,1587
UN No.: 3272
MDL No.: MFCD00009448
Formula:
C₇H₁₄O₂ Boiling Pt: 144…146 °C Melting Pt: –90 °C Density: 0.87 Flash Pt: 38 °C (101 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009448 CAS Number: 590-01-2 EINECS: 209-669-5 UN: 1914 ADR: 3,III Merck Index: 14,1587 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...