Tetrabromophenol blue
Supplier: Thermo Scientific Chemicals
Synonyms:
4,5,6,7-Tetrabromo-3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(4,5,6,7-Tetrabromo-3H-2,1-benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide
B20123-03
B20123-06
B20123-14
CAAAB20123-03EA
46.45
CAD
CAAAB20123-03
CAAAB20123-06
CAAAB20123-14
Tetrabromophenol blue
Tetrabromophenol blue
Phase transfer catalyst
CAS No.: 4430-25-5
Molecular Formula: C19H6Br8O5S
Formula Weight: 985.55
Melting Point: 203-205° dec.
Beilstein Reference: 378438
MDL No.: MFCD00005876
Formula:
C₁₉H₆Br₈O₅S MW: 985.55 g/mol Melting Pt: 203…205 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005876 CAS Number: 4430-25-5 EINECS: 224-622-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...