1-(4-Hydroxyphenyl)piperazine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Piperazin-1-yl)phenol, p-(1-Piperazinyl)phenol, 4-(1-Piperazinyl)phenol

B20122-18 B20122-30
CAAAB20122-18EA 317.58 CAD
CAAAB20122-18 CAAAB20122-30
1-(4-Hydroxyphenyl)piperazine 98%
1-(4-Hydroxyphenyl)piperazine

Purity: 98% CAS No.: 56621-48-8 Molecular Formula: C10H14N2O Formula Weight: 178.24 Melting Point: 218-224° Beilstein Reference: 150964 UN No.: 3263 MDL No.: MFCD00066156

Formula: C₁₀H₁₄N₂O
MW: 178.23 g/mol
Melting Pt: 218…224 °C
Storage Temperature: Ambient
MDL Number: MFCD00066156
CAS Number: 56621-48-8
EINECS: 260-289-6
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR