4-Bromo-2-fluorobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B20118-06 B20118-14
CAAAB20118-06EA 141.57 CAD
CAAAB20118-06 CAAAB20118-14
4-Bromo-2-fluorobenzoic acid 98%
4-Bromo-2-fluorobenzoic acid

Purity: 98% CAS No.: 112704-79-7 Molecular Formula: C7H4BrFO2 Formula Weight: 219.01 Melting Point: 210-213° Beilstein Reference: 6323931

Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
Melting Pt: 210…213 °C
Storage Temperature: Ambient
MDL Number: MFCD00143263
CAS Number: 112704-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR