sec-Pentylbenzene tech. 85%, Technical Grade
Supplier: Thermo Scientific Chemicals
B20114-03
B20114-06
CAAAB20114-03EA
62.25
CAD
CAAAB20114-03
CAAAB20114-06
CAAAB20114-14
sec-Pentylbenzene tech. 85%, Technical Grade
sec-Pentylbenzene
Purity: 85%
CAS No.: 2719-52-0
Molecular Formula: C11H16
Formula Weight: 148.25
MDL No.: MFCD00027145
Formula:
C₆H₅CH(CH₃)(CH₂)₂CH₃ Boiling Pt: 199 °C (1013 hPa) Melting Pt: –48 °C Density: 0.86 Storage Temperature: Ambient |
MDL Number:
MFCD00027145 CAS Number: 2719-52-0 EINECS: 220-324-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...