2,2,3,3,4,4,5,5-Octafluoro-1-pentanol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H,1H,5H-Octafluoro-1-pentanol

B20108-14 B20108-22
CAAAAB20108-36EA 334.55 CAD
CAAAAB20108-36 CAAAB20108-14 CAAAB20108-22
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol 98%
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol

Purity: 98% CAS No.: 355-80-6 Molecular Formula: F2CH(CF2)3CH2OH Formula Weight: 232.07 Flash Point: 75°(167°F) Refractive Index: 1.3180 Melting Point: -50° Boiling Point: 141-142° Beilstein Reference: 1773494 Merck Reference: 14,6746 MDL No.: MFCD00039631

Formula: C₅H₄F₈O
MW: 232.07 g/mol
Boiling Pt: 141
Melting Pt: < –50 °C
Density: 1.667
Flash Pt: 75 °C (167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039631
CAS Number: 355-80-6
EINECS: 206-593-4
Merck Index: 14,6746

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR