Benzyl mercaptan 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B20081-14
B20081-22
B20081-36
CAAAAB20081-14EA
47.87
CAD
CAAAAB20081-14
CAAAB20081-22
CAAAB20081-36
Benzyl mercaptan 99%
Benzyl mercaptan
Purity: 99%
CAS No.: 100-53-8
Molecular Formula: C7H8S
Formula Weight: 124.21
Flash Point: 70°(158°F)
Physical Form: Liquid
Density: 1.053
Refractive Index: 1.5760
Sensitivity: Air Sensitive
Appearance: Colorless to pale yellow
Odor: Stench
Melting Point: -30°
Boiling Point: 194-195°
Beilstein Reference: 605864
Merck Reference: 14,9322
UN No.: 2810
MDL No.: MFCD00004867
Formula:
C₇H₈S Boiling Pt: 194…195 °C Melting Pt: –30 °C Density: 1.053 Flash Pt: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004867 CAS Number: 100-53-8 EINECS: 202-862-5 UN: 2810 ADR: 6.1,III Merck Index: 14,9322 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...