2-Bromo-1-butene 97%

Supplier: Thermo Scientific Chemicals
Danger

B20067-06 B20067-14
CAAAB20067-06EA 510.34 CAD
CAAAB20067-06 CAAAB20067-14
2-Bromo-1-butene 97%
2-Bromo-1-butene

Purity: 97% CAS No.: 23074-36-4 Molecular Formula: CH3CH2CBr=CH2 Formula Weight: 135.00 Storage Temperature: -30°C to -10°C

Formula: C₄H₇Br
Boiling Pt: 88 °C (1013 hPa)
Melting Pt: –133.4 °C
Density: 1.315 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00039178
CAS Number: 23074-36-4
EINECS: 245-412-3
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR