Deoxycholic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

B20061-14 B20061-22
CAAAAB20061-36EA 536.85 CAD
CAAAAB20061-36 CAAAB20061-14 CAAAB20061-22
Deoxycholic acid 99%
Deoxycholic acid

Purity: 99% CAS No.: 83-44-3 Molecular Formula: C24H40O4 Formula Weight: 392.58 Melting Point: 172-176° Beilstein Reference: 3219882 Merck Reference: 14,2899 Optical Rotation: +54° (c=2 in ethanol) MDL No.: MFCD00003673

Formula: C₂₄H₄₀O₄
Boiling Pt: 504 °C (1013 hPa)
Melting Pt: 172…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00003673
CAS Number: 83-44-3
EINECS: 201-478-5
Merck Index: 14,2899

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR