1,4-Hexadiene cis- and trans-mixture 99%

Supplier: Thermo Scientific Chemicals
Danger

B20060-06 B20060-14
CAAAB20060-06EA 130.67 CAD
CAAAB20060-06 CAAAB20060-14
1,4-Hexadiene cis- and trans-mixture 99%
1,4-Hexadiene

Purity: 99% CAS No.: 592-45-0 Molecular Formula: CH3CH=CHCH2CH=CH2 Formula Weight: 82.15 Storage Temperature: 2°C to 8°C Flash Point: -26°(-15°F) Refractive Index: 1.4110 Odor: Characteristic Boiling Point: 64-66° Beilstein Reference: 1731889 UN No.: 2458 MDL No.: MFCD00009298

Formula: C₆H₁₀
Boiling Pt: 64…66 °C
Density: 0.710
Flash Pt: -26 °C(-15 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009298
CAS Number: 592-45-0
EINECS: 209-756-8
UN: 2458
ADR: "3",II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR