Triamterene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,4,7-Triamino-6-phenylpteridine, 6-Phenyl-2,4,7-triaminopteridine
B20044-06
B20044-14
B20044-22
CAAAB20044-06EA
66.52
CAD
CAAAB20044-06
CAAAB20044-14
CAAAB20044-22
Triamterene 98%
Triamterene
Weak diuretic with potassium sparing properties that blocks sodium reuptake in the kidneys
Purity: 98%
CAS No.: 396-01-0
Molecular Formula: C12H11N7
Formula Weight: 253.27
Melting Point:
Formula:
C₁₂H₁₁N₇ Boiling Pt: 573 °C (1013 hPa) Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006708 CAS Number: 396-01-0 EINECS: 206-904-3 Merck Index: 14,9599 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...