6-Hydroxy-m-toluic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B20040-06 B20040-14
CAAAB20040-06EA 93.54 CAD
CAAAB20040-06 CAAAB20040-14
6-Hydroxy-m-toluic acid 98%
6-Hydroxy-m-toluic acid

Purity: 98% CAS No.: 89-56-5 Molecular Formula: C8H8O3 Formula Weight: 152.15 Odor: Odorless Melting Point: 151-154° MDL No.: MFCD00002461

Formula: C₈H₈O₃
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 151…154 °C
Density: 1.31 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002461
CAS Number: 89-56-5
EINECS: 201-918-6
Merck Index: 12,02650

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR