4-(Trifluoromethoxy)phenyl isocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Isocyanato-4-(trifluoromethoxy)benzene
B20033-14
B20033-03
B20033-06
CAAAAB20033-14EA
1270.32
CAD
CAAAAB20033-14
CAAAB20033-03
CAAAB20033-06
4-(Trifluoromethoxy)phenyl isocyanate 97%
4-(Trifluoromethoxy)phenyl isocyanate
Purity: 97%
CAS No.: 35037-73-1
Molecular Formula: C8H4F3NO2
Formula Weight: 203.12
Storage Temperature: 2°C to 8°C
Flash Point: 70°(158°F)
Refractive Index: 1.4580
Melting Point: -29°
Boiling Point: 78°/22mm
Beilstein Reference: 977669
UN No.: 2206
MDL No.: MFCD00035702
Formula:
C₈H₄F₃NO₂ MW: 203.12 Boiling Pt: 78 Melting Pt: –29 °C Density: 1.346 Flash Pt: 70 °C (158 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00035702 CAS Number: 35037-73-1 EINECS: 252-328-0 UN: 3390 ADR: 6.1 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...