2,4-Dimethyl-1-pentene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B20002-09
B20002-88
CAAAB20002-09EA
734.7
CAD
CAAAB20002-09
CAAAB20002-88
2,4-Dimethyl-1-pentene 99%
2,4-Dimethyl-1-pentene
Purity: 99%
CAS No.: 2213-32-3
Molecular Formula: C7H14
Formula Weight: 98.19
Flash Point: -16°(-27°F)
Refractive Index: 1.3995
Boiling Point: 81°
UN No.: 3295
MDL No.: MFCD00039854
Formula:
C₇H₁₄ MW: 98.19 g/mol Boiling Pt: 81 °C Melting Pt: –124.1 °C Density: 0.69 Flash Pt: –16 °C (–27 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039854 CAS Number: 2213-32-3 EINECS: 218-665-2 UN: 3295 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...