3-Methyl-2-pentanone 98+%

Supplier: Thermo Scientific Chemicals
Danger

A19999-14 A19999-09 A19999-18
CAAAAA19999-14EA 314.42 CAD
CAAAAA19999-14 CAAAA19999-09 CAAAA19999-18
3-Methyl-2-pentanone 98+%
3-Methyl-2-pentanone

Purity: 98+% CAS No.: 565-61-7 Molecular Formula: C6H12O Formula Weight: 100.16 Flash Point: 12°(53°F) Refractive Index: 1.4 00 Boiling Point: 118-119° Beilstein Reference: 635791 UN No.: 1224 MDL No.: MFCD00009336

Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 118…119 °C
Density: 0.815
Flash Pt: 12 °C (53 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009336
CAS Number: 565-61-7
EINECS: 209-282-1
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR