Ethyl-2-nitrobenzoate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Ethyl 2-nitrobenzoate, 2-Nitrobenzoic acid ethyl ester
A19994-14
A19994-22
CAAAA19994-14EA
100.65
CAD
CAAAA19994-14
CAAAA19994-22
Ethyl-2-nitrobenzoate 97%
Ethyl 2-nitrobenzoate
Purity: 97%
CAS No.: 610-34-4
Molecular Formula: C9H9NO4
Formula Weight: 195.18
Melting Point: 26-30°
Boiling Point: 172-174°/18mm
Merck Reference: 14,3832
MDL No.: MFCD00014696
Formula:
C₉H₉NO₄ MW: 195.17 Boiling Pt: 172…174 °C (18 mmHg) Melting Pt: 26…30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014696 CAS Number: 610-34-4 EINECS: 210-220-0 Merck Index: 14,3832 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...