2,3-Dimethylanisole 97%

Supplier: Thermo Scientific Chemicals
Warning

A19992-09 A19992-18
CAAAA19992-09EA 82.63 CAD
CAAAA19992-09 CAAAA19992-18
2,3-Dimethylanisole 97%
2,3-Dimethylanisole

Purity: 97% CAS No.: 2944-49-2 Molecular Formula: C9H12O Formula Weight: 136.19 Flash Point: 67°(152°F) Refractive Index: 1.5200 Melting Point: 29° Boiling Point: 195° Beilstein Reference: 1859736 MDL No.: MFCD00008376

Formula: C₉H₁₂O
Boiling Pt: 195 °C
Melting Pt: 29 °C
Density: 0.984
Flash Pt: 67 °C(152 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008376
CAS Number: 2944-49-2
EINECS: 220-948-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR