3,5-Dichlorophenyl isocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A19991-06
A19991-14
CAAAA19991-06EA
122.45
CAD
CAAAA19991-06
CAAAA19991-14
3,5-Dichlorophenyl isocyanate 97%
3,5-Dichlorophenyl isocyanate
Purity: 97%
CAS No.: 34893-92-0
Molecular Formula: C7H3Cl2NO
Formula Weight: 188.01
Flash Point: 134°(273°F)
Melting Point: 28-32°
Boiling Point: 242-244°
Beilstein Reference: 510282
UN No.: 2250
MDL No.: MFCD00013859
Formula:
C₇H₃Cl₂NO Boiling Pt: 242…244 °C Melting Pt: 28…32 °C Density: 1.380 Flash Pt: 134 °C (273 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013859 CAS Number: 34893-92-0 EINECS: 252-276-9 UN: 2250 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...