Triisopropanolamine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,1',1''-Nitrilotripropan-2-ol, 1,1',1"-Nitrilotri-2-propanol, Tris(2-hydroxypropyl)amine

A19966-0C A19966-0E A19966-36
CAAAA19966-0CEA 548.26 CAD
CAAAA19966-0C CAAAA19966-0E CAAAA19966-36
Triisopropanolamine 95%
Triisopropanolamine

Purity: 95% CAS No.: 122-20-3 Molecular Formula: C9H21NO3 Formula Weight: 191.27 Flash Point: 110°(230°F) Odor: Amine-like Boiling Point: 189-191°/23mm Beilstein Reference: 1071570 MDL No.: MFCD00004533

Formula: C₉H₂₁NO₃
MW: 191.27 g/mol
Boiling Pt: 189…191 °C (23 mmHg)
Melting Pt: 48…52 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004533
CAS Number: 122-20-3
EINECS: 204-528-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR