4-Pentylbenzenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Amylbenzenesulfonyl chloride, 4-Pentylbenzene-1-sulfonyl chloride, 4-Pentylbenzene-1-sulphonyl chloride, 4-Amylbenzenesulphonyl chloride, 4-Amylbenzene-1-sulphonyl chloride, 4-Amylbenzene-1-sulfonyl chloride

A19929-09 A19929-88
CAAAA19929-09EA 141.41 CAD
CAAAA19929-09 CAAAA19929-88
4-Pentylbenzenesulfonyl chloride 97%
4-Pentylbenzenesulphonyl chloride

Purity: 97% CAS No.: 73948-18-2 Molecular Formula: C11H15ClO2S Formula Weight: 246.76 Melting Point: 39-44° UN No.: 3261 MDL No.: MFCD00173759

Formula: C₁₁H₁₅ClO₂S
Melting Pt: 39…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00173759
CAS Number: 73948-18-2
UN: 3261
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR