N-Aminophthalimide 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19914-06
A19914-09
A19914-18
CAAAAA19914-06EA
97.17
CAD
CAAAAA19914-06
CAAAA19914-09
CAAAA19914-18
N-Aminophthalimide 94%
N-Aminophthalimide
Purity: 94%
CAS No.: 1875-48-5
Molecular Formula: C8H6N2O2
Formula Weight: 162.15
Storage Temperature: 2°C to 8°C
Melting Point: ca 195° dec.
Beilstein Reference: 383756
MDL No.: MFCD00005895
Formula:
C₈H₆N₂O₂ Boiling Pt: 391 °C (1013 hPa) Melting Pt: ∼ 195 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005895 CAS Number: 1875-48-5 EINECS: 217-505-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...