2'-Fluoro-4'-hydroxyacetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(2-Fluoro-4-hydroxyphenyl)ethanone
A19884-06
A19884-14
CAAAAA19884-03EA
67.29
CAD
CAAAAA19884-03
CAAAA19884-06
CAAAA19884-14
2'-Fluoro-4'-hydroxyacetophenone 97%
2'-Fluoro-4'-hydroxyacetophenone
Purity: 97%
CAS No.: 98619-07-9
Molecular Formula: C8H7FO2
Formula Weight: 154.14
Melting Point: 121-123°
Beilstein Reference: 696642
Formula:
C₈H₇FO₂ MW: 154.14 g/mol Melting Pt: 121…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD00142713 CAS Number: 98619-07-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...