3-Nonanone 98%
Supplier: Thermo Scientific Chemicals
Warning
|
A19878-14
A19878-22
CAAAA19878-14EA
78.37
CAD
CAAAA19878-14
CAAAA19878-22
3-Nonanone 98%
3-Nonanone
Purity: 98%
CAS No.: 925-78-0
Molecular Formula: C9H18O
Formula Weight: 142.24
Flash Point: 63°(145°F)
Refractive Index: 1.4200
Melting Point: -8°
Boiling Point: 187-188°
Beilstein Reference: 1700447
MDL No.: MFCD00009541
Formula:
C₉H₁₈O MW: 142.24 g/mol Boiling Pt: 188 Melting Pt: -8 Density: 0.825 Flash Pt: 63 °C (145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009541 CAS Number: 925-78-0 EINECS: 213-125-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...