5-Iodoanthranilic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-5-iodobenzoic acid

A19838-06 A19838-14
CAAAA19838-06EA 23.54 CAD
CAAAA19838-06 CAAAA19838-14
5-Iodoanthranilic acid 98%
5-Iodoanthranilic acid

Purity: 98% CAS No.: 5326-47-6 Molecular Formula: C7H6INO2 Formula Weight: 263.03 Melting Point: ca 220° dec. Beilstein Reference: 639029

Formula: C₇H₆INO₂
MW: 263.03 g/mol
Boiling Pt: 360
Melting Pt: 234 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007849
CAS Number: 5326-47-6
EINECS: 226-205-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR