Cyclohexanone oxime 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A19820-22
A19820-36
CAAAA19820-22EA
41.71
CAD
CAAAA19820-22
CAAAA19820-36
Cyclohexanone oxime 97%
Cyclohexanone oxime
Purity: 97%
CAS No.: 100-64-1
Molecular Formula: C6H11NO
Formula Weight: 113.16
Flash Point: 103°(217°F)
Melting Point: 87-91°
Boiling Point: 206-210°
Beilstein Reference: 1616769
MDL No.: MFCD00001660
Formula:
C₆H₁₁NO MW: 113.16 g/mol Boiling Pt: 206…210 °C Melting Pt: 87…91 °C Density: 1.13 g/cm³ (20 °C) Flash Pt: 103 °C (217 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001660 CAS Number: 100-64-1 EINECS: 202-874-0 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...