Ethyl-2-chloroacetoacetate 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A19810-22
A19810-36
CAAAA19810-22EA
60.36
CAD
CAAAA19810-22
CAAAA19810-36
Ethyl-2-chloroacetoacetate 96%
Ethyl 2-chloroacetoacetate
Purity: 96%
CAS No.: 609-15-4
Molecular Formula: C6H9ClO3
Formula Weight: 164.59
Flash Point: 82°(180°F)
Refractive Index: 1.4425
Melting Point:
Formula:
C₆H₉ClO₃ Boiling Pt: 200 °C Melting Pt: < –80 °C Density: 1.182 Flash Pt: 54 °C(180 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009141 CAS Number: 609-15-4 EINECS: 210-180-4 UN: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...