3,5-Diiodosalicylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-3,5-diiodobenzoic acid

A19786-22 A19786-36
CAAAA19786-14EA 118.12 CAD
CAAAA19786-14 CAAAA19786-22 CAAAA19786-36
3,5-Diiodosalicylic acid 97%
3,5-Diiodosalicylic acid

Purity: 97% CAS No.: 133-91-5 Molecular Formula: C7H4I2O3 Formula Weight: 389.91 Melting Point: 228-234° Beilstein Reference: 2615358 Merck Reference: 14,3190 MDL No.: MFCD00002444

Formula: C₇H₄I₂O₃
MW: 389.92 g/mol
Boiling Pt: 381 °C (1013 hPa)
Melting Pt: 228…234 °C
Storage Temperature: Ambient
MDL Number: MFCD00002444
CAS Number: 133-91-5
EINECS: 205-124-0
Merck Index: 14,3190

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR