1-Bromo-3-chloro-4-fluorobenzene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromo-2-chloro-1-fluorobenzene, 4-Bromo-2-chlorofluorobenzene
A19782-22
A19782-14
CAAAAA19782-22EA
1038.06
CAD
CAAAAA19782-22
CAAAA19782-14
1-Bromo-3-chloro-4-fluorobenzene 98+%
1-Bromo-3-chloro-4-fluorobenzene
Purity: 98+%
CAS No.: 60811-21-4
Molecular Formula: C6H3BrClF
Formula Weight: 209.45
Flash Point: 88°(190°F)
Refractive Index: 1.5540
Boiling Point: 188-190°
Beilstein Reference: 2206652
UN No.: 3082
MDL No.: MFCD00051794
Formula:
C₆H₃BrClF Boiling Pt: 188…190 °C Density: 1.727 Flash Pt: 88 °C (190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051794 CAS Number: 60811-21-4 EINECS: 405-580-2 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...