2,4,6-Trifluorobenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19758-03
A19758-06
CAAAA19758-03EA
82.16
CAD
CAAAA19758-03
CAAAA19758-06
2,4,6-Trifluorobenzoic acid 98%
2,4,6-Trifluorobenzoic acid
Purity: 98%
CAS No.: 28314-80-9
Molecular Formula: C7H3F3O2
Formula Weight: 176.10
Melting Point: 140-142°
Beilstein Reference: 1958300
Formula:
C₇H₃F₃O₂ Boiling Pt: 237 °C (1013 hPa) Melting Pt: 140…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042398 CAS Number: 28314-80-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...