4-(Trifluoromethoxy)anisole 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19746-06
A19746-14
CAAAA19746-06EA
45.19
CAD
CAAAA19746-06
CAAAA19746-14
4-(Trifluoromethoxy)anisole 98+%
4-(Trifluoromethoxy)anisole
Purity: 98+%
CAS No.: 710-18-9
Molecular Formula: C8H7F3O2
Formula Weight: 192.14
Flash Point: 58°(136°F)
Refractive Index: 1.4320
Boiling Point: 163-165°
Beilstein Reference: 2556809
UN No.: 1993
MDL No.: MFCD00216942
Formula:
C₈H₇F₃O₂ Boiling Pt: 163…165 °C Density: 1.272 Flash Pt: 58 °C (136 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00216942 CAS Number: 710-18-9 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...