(±)-γ-Decalactone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Hexyldihydrofuran-2(3H)-one, Gamma-decalactone, (±)-Decan-4-olide
A19732-06
A19732-14
A19732-22
CAAAAA19732-06EA
47.24
CAD
CAAAAA19732-06
CAAAA19732-14
CAAAA19732-22
(±)-γ-Decalactone 98%
(±)-γ-Decalactone
Purity: 98%
CAS No.: 706-14-9
Molecular Formula: C10H18O2
Formula Weight: 170.25
Flash Point: 146°(294°F)
Refractive Index: 1.500
Boiling Point: 150-153°/15mm
Beilstein Reference: 117547
MDL No.: MFCD00005404
Formula:
C₁₀H₁₈O₂ MW: 170.25 g/mol Boiling Pt: 150…153 °C/15 mm Density: 0,948 Flash Pt: 146 °C(294 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005404 CAS Number: 706-14-9 EINECS: 211-892-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...