2-Fluoro-p-anisaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Fluoro-4-methoxybenzaldehyde, 2-Fluoro-4-anisaldehyde
A19726-06
A19726-14
CAAAA19726-03EA
80.64
CAD
CAAAA19726-03
CAAAA19726-06
CAAAA19726-14
2-Fluoro-p-anisaldehyde 97%
2-Fluoro-p-anisaldehyde
Purity: 97%
CAS No.: 331-64-6
Molecular Formula: C8H7FO2
Formula Weight: 154.14
Melting Point: 42-47°
Beilstein Reference: 3237954
Formula:
C₈H₇FO₂ Melting Pt: 42…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236679 CAS Number: 331-64-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...