2,4-Dichloro-5-methylbenzenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

A19718-09 A19718-18 A19718-88
CAAAA19718-09EA 284.4 CAD
CAAAA19718-09 CAAAA19718-18 CAAAA19718-88
2,4-Dichloro-5-methylbenzenesulfonyl chloride 97%
2,4-Dichloro-5-methylbenzenesulphonyl chloride

Purity: 97% CAS No.: 28286-86-4 Molecular Formula: C7H5Cl3O2S Formula Weight: 259.54 Melting Point: 70-73° UN No.: 3261 MDL No.: MFCD00044016

Formula: C₇H₅Cl₃O₂S
Melting Pt: 70…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00044016
CAS Number: 28286-86-4
EINECS: 248-938-1
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR