2,4-Dihydroxy-3-nitro-6-picoline 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,4-Dihydroxy-6-methyl-3-nitropyridine, 4-Hydroxy-6-methyl-3-nitropyridin-2(1H)-one, 4-Hydroxy-6-methyl-3-nitro-2-pyridone

A19690-14 A19690-22
CAAAA19690-06EA 106.27 CAD
CAAAA19690-06 CAAAA19690-14 CAAAA19690-22
2,4-Dihydroxy-3-nitro-6-picoline 97%
2,4-Dihydroxy-3-nitro-6-picoline

Purity: 97% CAS No.: 4966-90-9 Molecular Formula: C6H6N2O4 Formula Weight: 170.13 Melting Point: 250° dec. Beilstein Reference: 1529425

Formula: C₆H₆N₂O₄
MW: 170.12 g/mol
Melting Pt: >250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00167612
CAS Number: 4966-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR