1,3-Dimethyladamantane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19684-06
A19684-14
CAAAA19684-03EA
81.77
CAD
CAAAA19684-03
CAAAA19684-06
CAAAA19684-14
1,3-Dimethyladamantane 98%
1,3-Dimethyladamantane
Purity: 98%
CAS No.: 702-79-4
Molecular Formula: C12H20
Formula Weight: 164.29
Flash Point: 52°(126°F)
Refractive Index: 1.4780
Boiling Point: 201°
UN No.: 3295
MDL No.: MFCD00074755
Formula:
C₁₂H₂₀ Boiling Pt: 201 °C Melting Pt: –30 °C Density: 0.902 Flash Pt: 52 °C(126 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00074755 CAS Number: 702-79-4 EINECS: 211-870-8 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...