4-Fluoro-3-nitrobenzotrifluoride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19676-14
A19676-22
A19676-36
CAAAA19676-14EA
48.19
CAD
CAAAA19676-14
CAAAA19676-22
CAAAA19676-36
4-Fluoro-3-nitrobenzotrifluoride 97%
4-Fluoro-3-nitrobenzotrifluoride
Purity: 97%
CAS No.: 367-86-2
Molecular Formula: C7H3F4NO2
Formula Weight: 209.10
Flash Point: 90°(194°F)
Refractive Index: 1.4620
Boiling Point: 91-92°/15mm
Beilstein Reference: 1880508
UN No.: 2810
MDL No.: MFCD00007059
Formula:
C₇H₃F₄NO₂ Boiling Pt: 91…92 °C/15 mm Density: 1.494 Flash Pt: 90 °C (194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007059 CAS Number: 367-86-2 EINECS: 206-702-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...