1,3-Diphenyl-2-thiourea 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Thiocarbanilide, N,N'-Diphenylthiourea, N,N’-Diphenylthiourea

A19644-22 A19644-0E A19644-36
CAAAAA19644-22EA 41.87 CAD
CAAAAA19644-22 CAAAA19644-0E CAAAA19644-36
1,3-Diphenyl-2-thiourea 98%
1,3-Diphenyl-2-thiourea

Purity: 98% CAS No.: 102-08-9 Molecular Formula: C13H12N2S Formula Weight: 228.32 Melting Point: 150-154° Beilstein Reference: 644277 Merck Reference: 14,3333 UN No.: 2811 MDL No.: MFCD00004921

Formula: C₁₃H₁₂N₂S
MW: 228.32 g/mol
Boiling Pt: 365
Melting Pt: 150…154 °C
Density: 1.320
Storage Temperature: Ambient
MDL Number: MFCD00004921
CAS Number: 102-08-9
EINECS: 203-004-2
UN: 3077
ADR: 9 III
Merck Index: 14,3333

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR