(±)-γ-Hexalactone 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(±)-γ-Caprolactone, γ-Caprolactone, 5-Ethyldihydrofuran-2(3H)-one, (±)-Hexan-4-olide
A19642-14
A19642-22
CAAAA19642-14EA
48.03
CAD
CAAAA19642-14
CAAAA19642-22
(±)-γ-Hexalactone 98%
(±)-γ-Hexalactone
Purity: 98%
CAS No.: 695-06-7
Molecular Formula: C6H10O2
Formula Weight: 114.15
Flash Point: 98°(208°F)
Refractive Index: 1.4390
Melting Point: -18°
Boiling Point: 218-219°
Beilstein Reference: 107260
MDL No.: MFCD00005401
Formula:
C₆H₁₀O₂ MW: 114.14 g/mol Boiling Pt: 218…219 °C Melting Pt: –18 °C Density: 1.025 Flash Pt: 98 °C (208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005401 CAS Number: 695-06-7 EINECS: 211-778-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...