N,N-Diethylethylenediamine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A19636-22
A19636-36
CAAAA19636-22EA
51.82
CAD
CAAAA19636-22
CAAAA19636-36
N,N-Diethylethylenediamine 99%
N,N-Diethylethylenediamine
Purity: 99%
CAS No.: 100-36-7
Molecular Formula: (CH3CH2)2NCH2CH2NH2
Formula Weight: 116.21
Flash Point: 30°(86°F)
Refractive Index: 1.4360
Melting Point:
Formula:
C₆H₁₆N₂ Boiling Pt: 145…147 °C Melting Pt: < –70 °C Density: 0.825 Flash Pt: 30 °C(86 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008176 CAS Number: 100-36-7 EINECS: 202-844-7 UN: 2685 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...