3-Bromobenzhydrazide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19628-06
A19628-14
CAAAA19628-06EA
70.15
CAD
CAAAA19628-06
CAAAA19628-14
3-Bromobenzhydrazide 98+%
3-Bromobenzhydrazide
Purity: 98+%
CAS No.: 39115-96-3
Molecular Formula: C7H7BrN2O
Formula Weight: 215.05
Melting Point: 154-157°
Beilstein Reference: 1945814
Formula:
C₇H₇BrN₂O Boiling Pt: 366 °C (1013 hPa) Melting Pt: 154…157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007600 CAS Number: 39115-96-3 EINECS: 254-298-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...