1,3-Diamino-2-propanol 97%

Supplier: Thermo Scientific Chemicals
Warning

A19618-09 A19618-18
CAAAA19618-09EA 110.76 CAD
CAAAA19618-09 CAAAA19618-18
1,3-Diamino-2-propanol 97%
1,3-Diamino-2-propanol

Purity: 97% CAS No.: 616-29-5 Molecular Formula: C3H10N2O Formula Weight: 90.13 Flash Point: 110°(230°F) Melting Point: 40-45° Boiling Point: 235° Beilstein Reference: 741859 MDL No.: MFCD00008142

Formula: C₃H₁₀N₂O
Boiling Pt: 235 °C
Melting Pt: 40…45 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008142
CAS Number: 616-29-5
EINECS: 210-474-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR