Trimethylsilyl cyanide 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethylsilylcarbonitrile

A19598-06 A19598-14 A19598-22
CAAAA19598-06EA 91.48 CAD
CAAAA19598-06 CAAAA19598-14 CAAAA19598-22
Trimethylsilyl cyanide 97%
Trimethylsilyl cyanide

Purity: 97% CAS No.: 7677-24-9 Molecular Formula: C4H9NSi Formula Weight: 99.21 Flash Point: 1°(33°F) Physical Form: Liquid Density: 0.783 Refractive Index: 1.3910 Sensitivity: Moisture Sensitive Odor: Almond-like Melting Point: 11-12° Boiling Point: 118-119° Beilstein Reference: 1737612 UN No.: 3383 MDL No.: MFCD00001765

Formula: C₄H₉NSi
Boiling Pt: 118…119 °C
Melting Pt: 11…12 °C
Density: 0.783
Flash Pt: 1 °C (33 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001765
CAS Number: 7677-24-9
EINECS: 231-657-3
UN: 3383
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR