1-Bromo-3-iodobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

A19596-22 A19596-14
CAAAAA19596-22EA 529.3 CAD
CAAAAA19596-22 CAAAA19596-14
1-Bromo-3-iodobenzene 98%
1-Bromo-3-iodobenzene

Purity: 98% CAS No.: 591-18-4 Molecular Formula: C6H4BrI Formula Weight: 282.91 Flash Point: 110°(230°F) Refractive Index: 1.6620 Odor: Recognizable Melting Point: -9° Boiling Point: 252° Beilstein Reference: 1854384 MDL No.: MFCD00001043

Formula: C₆H₄BrI
Boiling Pt: 252 °C
Melting Pt: –9 °C
Density: 2.219
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001043
CAS Number: 591-18-4
EINECS: 209-703-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR