3-(Trifluoromethoxy)aniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19592-03
A19592-06
CAAAA19592-03EA
91.32
CAD
CAAAA19592-03
CAAAA19592-06
3-(Trifluoromethoxy)aniline 98%
3-(Trifluoromethoxy)aniline
Purity: 98%
CAS No.: 1535-73-5
Molecular Formula: C7H6F3NO
Formula Weight: 177.13
Flash Point: 85°(185°F)
Refractive Index: 1.4660
Odor: Characteristic
Boiling Point: 72-75°/8mm
Beilstein Reference: 776921
UN No.: 2810
MDL No.: MFCD00041511
Formula:
C₇H₆F₃NO Boiling Pt: 72…75 °C (8 mmHg) Density: 1.325 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041511 CAS Number: 1535-73-5 EINECS: 216-256-3 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...